Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCED1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281429.1 | NP_001268358.1 | 3 | 1516 | ||
NM_138371.2 | NP_612380.1 | 4 | 1984 | |||
XM_005269224.4 | XP_005269281.1 | 4 | 1681 | |||
XM_011538978.2 | XP_011537280.1 | 5 | 1849 | |||
XM_017020207.1 | XP_016875696.1 | 6 | 1906 | |||
XM_017020208.1 | XP_016875697.1 | 7 | 1983 | |||
XM_017020209.1 | XP_016875698.1 | 5 | 1738 | |||
XM_017020210.1 | XP_016875699.1 | 6 | 3655 | |||
XM_017020211.1 | XP_016875700.1 | 3 | 1935 | |||
XM_017020212.1 | XP_016875701.1 | 4 | 1678 | |||
XM_017020213.1 | XP_016875702.1 | 7 | 2051 | |||
XM_017020214.1 | XP_016875703.1 | 8 | 2167 | |||
XM_017020215.1 | XP_016875704.1 | 7 | 2022 | |||
XM_017020216.1 | XP_016875705.1 | 3 | 3226 | |||
GenBank mRNA | BC008360.1 | - | 2 | 1731 | 3610443 | |
BC016154.1 | - | 3 | 938 | 4040497 | ||
BC072670.1 | - | 4 | 1982 | |||
BM545125.1 | - | 1 | 168 |