Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCED1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281429.1 | NP_001268358.1 | 3 | 1368 | ||
NM_138371.2 | NP_612380.1 | 4 | 1836 | |||
XM_005269224.4 | XP_005269281.1 | 4 | 1533 | |||
XM_011538978.2 | XP_011537280.1 | 5 | 1701 | |||
XM_017020207.1 | XP_016875696.1 | 6 | 1758 | |||
XM_017020208.1 | XP_016875697.1 | 7 | 1835 | |||
XM_017020209.1 | XP_016875698.1 | 5 | 1590 | |||
XM_017020210.1 | XP_016875699.1 | 6 | 3507 | |||
XM_017020211.1 | XP_016875700.1 | 3 | 1787 | |||
XM_017020212.1 | XP_016875701.1 | 4 | 1530 | |||
XM_017020213.1 | XP_016875702.1 | 7 | 1903 | |||
XM_017020214.1 | XP_016875703.1 | 8 | 2019 | |||
XM_017020215.1 | XP_016875704.1 | 7 | 1874 | |||
XM_017020216.1 | XP_016875705.1 | 3 | 3078 | |||
GenBank mRNA | BC008360.1 | - | 2 | 1583 | 3610443 | |
BC016154.1 | - | 3 | 790 | 4040497 | ||
BC072670.1 | - | 4 | 1834 |