Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 10 | 1038 | ||
NM_145345.2 | NP_663320.2 | 14 | 1518 | |||
NM_183008.2 | NP_892120.2 | 15 | 1523 | |||
GenBank mRNA | AF370425.1 | - | 4 | 1312 | ||
AF520989.1 | - | 13 | 1227 | |||
AF521017.1 | - | 13 | 1228 | |||
AF521018.1 | - | 13 | 1215 | |||
AK074358.1 | - | 9 | 2032 | |||
AK090792.1 | - | 9 | 1997 | |||
AK093258.1 | - | 15 | 1467 | |||
AK097368.1 | - | 14 | 1616 | |||
AY007163.1 | - | 11 | 982 | |||
BC038106.1 | - | 13 | 2316 | 5269682 | ||
BX647774.1 | - | 13 | 2312 | |||
BX648412.1 | - | 12 | 2213 | |||
CR749413.1 | - | 13 | 2346 |