Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104587.1 | NP_001098057.1 | 4 | 1407 | ||
NM_001104588.1 | NP_001098058.1 | 4 | 1325 | |||
NM_001104589.1 | NP_001098059.1 | 3 | 1247 | |||
NM_001104590.1 | NP_001098060.1 | 4 | 1237 | |||
NM_152270.3 | NP_689483.3 | 2 | 1130 | |||
XM_005258068.3 | XP_005258125.1 | 2 | 1235 | |||
XM_006722171.3 | XP_006722234.1 | 4 | 1218 | |||
XM_011525465.2 | XP_011523767.1 | 3 | 1440 | |||
XM_017025299.1 | XP_016880788.1 | 4 | 1558 | |||
GenBank mRNA | AK074184.1 | - | 2 | 994 | ||
AK092241.1 | - | 4 | 1160 | |||
AL831964.2 | - | 2 | 1030 | |||
BC052586.1 | - | 2 | 1036 | 6258140 |