Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASL10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033315.3 | NP_201572.1 | 4 | 1868 | ||
XM_017025300.1 | XP_016880789.1 | 5 | 2762 | |||
XM_017025301.1 | XP_016880790.1 | 3 | 2502 | |||
GenBank mRNA | AK122652.1 | - | 4 | 1866 | ||
BC041133.1 | - | 4 | 1788 | 5762715 | ||
DA250459.1 | - | 1 | 465 |