Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEPDC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077242.1 | NP_001070710.1 | 6 | 1101 | ||
NM_139160.2 | NP_631899.2 | 6 | 1340 | |||
GenBank mRNA | AF447588.1 | - | 6 | 1340 | ||
AJ245600.1 | - | 6 | 1056 | |||
BC030970.1 | - | 6 | 1101 | 4133262 |