Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BOC SIRNA Assays ›
Gene Symbol:
BOC
Gene Name:
BOC cell adhesion associated, oncogene regulated
Gene Aliases:
CDON2
Chromosome Location:
Chr.3: 113211003 - 113287463 on Build GRCh38
UniGene ID:
Hs.591318
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40723
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BOC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301861.1
NP_001288790.1
15
2793
NM_033254.3
NP_150279.1
15
3141
XM_005247891.2
XP_005247948.1
15
3131
XM_005247892.2
XP_005247949.1
15
3127
XM_005247897.2
XP_005247954.1
14
2858
XM_011513305.2
XP_011511607.1
15
3128
XM_017007441.1
XP_016862930.1
15
3140
XM_017007442.1
XP_016862931.1
15
3136
XM_017007443.1
XP_016862932.1
15
2928
XM_017007444.1
XP_016862933.1
15
2720
XM_017007445.1
XP_016862934.1
15
2793
XM_017007446.1
XP_016862935.1
15
2760
XM_017007447.1
XP_016862936.1
15
2751
XM_017007448.1
XP_016862937.1
15
2672
XM_017007449.1
XP_016862938.1
15
2879
XM_017007450.1
XP_016862939.1
15
3137
XM_017007451.1
XP_016862940.1
15
3270
XM_017007452.1
XP_016862941.1
13
2303
XM_017007453.1
XP_016862942.1
13
2294
XM_017007454.1
XP_016862943.1
13
2679
GenBank mRNA
AK074556.1
-
8
1475
AK090455.1
-
9
4780
AY027658.1
-
14
2552
AY358328.1
-
16
2762
BC032071.1
-
3
236
BC075829.1
-
15
3013
BC136390.1
-
14
2578
9020377
BX647184.1
-
9
3154
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
smoothened signaling pathway
axon guidance
positive regulation of myoblast differentiation
positive regulation of muscle cell differentiation
mitophagy in response to mitochondrial depolarization
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0