Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020818.2 | NP_001018654.1 | 2 | 1241 | ||
NM_001020819.2 | NP_001018655.1 | 3 | 1021 | |||
NM_001020820.2 | NP_001018656.1 | 3 | 1138 | |||
NM_001020821.2 | NP_001018657.1 | 3 | 1086 | |||
NM_001290188.1 | NP_001277117.1 | 3 | 1202 | |||
NM_001290189.1 | NP_001277118.1 | 3 | 1472 | |||
NM_001290190.1 | NP_001277119.1 | 3 | 993 | |||
NM_001290191.1 | NP_001277120.1 | 3 | 945 | |||
NM_001290192.1 | NP_001277121.1 | 3 | 962 | |||
NM_001290193.1 | NP_001277122.1 | 3 | 1023 | |||
NM_001290194.1 | NP_001277123.1 | 2 | 872 | |||
NM_138373.4 | NP_612382.1 | 3 | 957 | |||
GenBank mRNA | AF087882.1 | - | 3 | 1070 | ||
AK027693.1 | - | 1 | 579 | |||
AK075276.1 | - | 3 | 939 | |||
AK093999.1 | - | 4 | 883 | |||
AK294400.1 | - | 4 | 828 | |||
AK316566.1 | - | 3 | 935 | |||
AL524210.3 | - | 2 | 835 | |||
AX775819.1 | - | 3 | 937 | |||
AY037147.2 | - | 3 | 960 | |||
AY358582.1 | - | 3 | 921 | |||
BC013995.2 | - | 2 | 1193 | 3627858 | ||
BC095412.1 | - | 3 | 939 | 30527914 |