Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031732.2 | NP_001026902.1 | 2 | 409 | ||
NM_133370.2 | NP_588611.2 | 2 | 409 | |||
XM_005265706.2 | XP_005265763.1 | 2 | 409 | |||
XM_005265707.2 | XP_005265764.1 | 2 | 393 | |||
XM_005265708.2 | XP_005265765.1 | 2 | 409 | |||
XM_017008832.1 | XP_016864321.1 | 2 | 393 | |||
XM_017008833.1 | XP_016864322.1 | 2 | 393 | |||
GenBank mRNA | AK056430.1 | - | 2 | 394 | ||
BC041119.1 | - | 2 | 339 | 5541053 | ||
BC053863.1 | - | 2 | 74 | 4933750 | ||
DA191646.1 | - | 2 | 409 |