Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 10 | 3058 | ||
NM_194278.3 | NP_919254.2 | 10 | 3437 | |||
XM_005268204.1 | XP_005268261.1 | 10 | 3437 | |||
XM_005268205.1 | XP_005268262.1 | 10 | 2993 | |||
XM_005268206.1 | XP_005268263.1 | 11 | 3210 | |||
GenBank mRNA | AK127785.1 | - | 11 | 3436 | ||
BC003121.1 | - | 10 | 2230 | |||
BC006451.1 | - | 10 | 1892 | |||
BC006511.2 | - | 10 | 2688 | |||
BC009202.2 | - | 11 | 3058 | |||
BC015668.2 | - | 10 | 3071 | |||
BC025330.1 | - | 10 | 2688 |