Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153369.2 | NP_699200.2 | 4 | 944 | ||
XM_017011483.1 | XP_016866972.1 | 3 | 827 | |||
GenBank mRNA | AB067506.1 | - | 1 | 179 | ||
AK093136.1 | - | 4 | 903 | |||
AK292735.1 | - | 4 | 873 | |||
AK315016.1 | - | 4 | 866 | |||
BC036115.1 | - | 4 | 853 | 5298999 |