Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 8 | 1428 | ||
GenBank mRNA | AB073391.1 | - | 9 | 1303 | ||
AF261758.1 | - | 8 | 1398 | |||
AK298414.1 | - | 3 | 657 | |||
AK302774.1 | - | 8 | 1259 | |||
AK304302.1 | - | 5 | 716 | |||
AK304421.1 | - | 4 | 555 | |||
AK304437.1 | - | 8 | 1145 | |||
AK316419.1 | - | 8 | 1190 | |||
AK316427.1 | - | 8 | 1129 | |||
BC004375.1 | - | 8 | 1334 | 3638639 | ||
BC011669.2 | - | 8 | 1361 | 4130396 | ||
D13643.3 | - | 8 | 1411 |