Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHCR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014762.3 | NP_055577.1 | 8 | 1438 | ||
GenBank mRNA | AB073391.1 | - | 9 | 1313 | ||
AF261758.1 | - | 8 | 1408 | |||
AK298414.1 | - | 3 | 667 | |||
AK302774.1 | - | 8 | 1269 | |||
AK304302.1 | - | 5 | 726 | |||
AK304421.1 | - | 4 | 565 | |||
AK304437.1 | - | 8 | 1155 | |||
AK316419.1 | - | 8 | 1200 | |||
AK316427.1 | - | 8 | 1139 | |||
BC004375.1 | - | 8 | 1344 | 3638639 | ||
BC011669.2 | - | 8 | 1371 | 4130396 | ||
D13643.3 | - | 8 | 1421 |