Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143981.1 | NP_001137453.1 | 9 | 1219 | ||
NM_001143982.1 | NP_001137454.1 | 9 | 1216 | |||
NM_001143983.2 | NP_001137455.2 | 7 | 979 | |||
NM_145234.3 | NP_660277.2 | 9 | 1064 | |||
XM_005262221.1 | XP_005262278.1 | 9 | 1036 | |||
XM_005262222.3 | XP_005262279.1 | 9 | 1033 | |||
XM_005262223.1 | XP_005262280.1 | 9 | 1013 | |||
XM_005262224.1 | XP_005262281.1 | 9 | 1032 | |||
XM_017029959.1 | XP_016885448.1 | 9 | 1036 | |||
GenBank mRNA | AB209548.1 | - | 8 | 809 | ||
AK092245.1 | - | 6 | 646 | |||
AK293106.1 | - | 9 | 966 | |||
AK297563.1 | - | 7 | 740 | |||
AK312270.1 | - | 9 | 977 | |||
AY608914.1 | - | 8 | 893 | |||
BC002909.2 | - | 9 | 981 | 3942645 |