Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031733.2 | NP_001026903.2 | 3 | 1043 | ||
NM_001286694.1 | NP_001273623.1 | 3 | 1153 | |||
NM_001286695.1 | NP_001273624.1 | 2 | 1012 | |||
NM_033429.2 | NP_219501.2 | 4 | 1184 | |||
GenBank mRNA | AF308287.1 | - | 4 | 632 | ||
AK092628.1 | - | 3 | 265 | |||
AK296804.1 | - | 4 | 406 | |||
BC009516.2 | - | 3 | 360 | |||
BX640702.1 | - | 2 | 201 | |||
DB204573.1 | - | 4 | 408 |