Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153603.3 | NP_705831.1 | 10, 11 | 1603 | ||
XM_017023870.1 | XP_016879359.1 | 10, 11 | 1646 | |||
GenBank mRNA | AF070568.1 | - | 2, 3 | 175 | ||
AK093989.1 | - | 7, 8 | 959 | |||
AY358632.1 | - | 5, 6 | 640 | |||
BC000549.2 | - | 10, 11 | 1558 | 3163316 | ||
BC037563.1 | - | 10, 11 | 1592 | 5393265 | ||
KU178837.1 | - | 10, 11 | 1406 |