Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122853.2 | NP_001116325.1 | 7 | 1487 | ||
NM_133371.4 | NP_588612.2 | 7 | 1139 | |||
GenBank mRNA | AF052497.1 | - | 2 | 947 | ||
AJ300586.1 | - | 7 | 1102 | |||
AJ300587.1 | - | 7 | 844 |