Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001361.4 | NP_001352.2 | 3, 4 | 463 | ||
XM_005255827.3 | XP_005255884.1 | 3, 4 | 768 | |||
XM_017022990.1 | XP_016878479.1 | 3, 4 | 543 | |||
GenBank mRNA | AK097300.1 | - | 1 | 1409 | ||
AK292293.1 | - | 4, 5 | 619 | |||
AU130529.1 | - | 4, 5 | 463 | |||
BC065245.1 | - | 3, 4 | 443 | 6064723 | ||
BI253747.1 | - | 3, 4 | 447 | |||
BM795077.1 | - | 2, 3 | 238 | |||
M94065.1 | - | 4, 5 | 445 |