Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122853.2 | NP_001116325.1 | 3 | 711 | ||
NM_133371.4 | NP_588612.2 | 3 | 363 | |||
GenBank mRNA | AF480443.1 | - | 3 | 215 | ||
AJ277961.1 | - | 2 | 690 | |||
AJ300586.1 | - | 3 | 326 | |||
AJ300587.1 | - | 3 | 206 | |||
AK313873.1 | - | 3 | 367 | |||
BC069380.1 | - | 3 | 193 | 7262195 | ||
BC069553.1 | - | 3 | 193 | 7262207 | ||
BC074870.2 | - | 3 | 149 | 30915232 | ||
BC074871.2 | - | 3 | 149 | 30915469 | ||
DB069077.1 | - | 3 | 367 | |||
DB277496.1 | - | 3 | 366 |