Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM58A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130997.2 | NP_001124469.1 | 2 | 282 | ||
NM_152274.4 | NP_689487.2 | 2 | 282 | |||
XM_005277920.4 | XP_005277977.1 | 2 | 270 | |||
XM_005277921.4 | XP_005277978.1 | 2 | 270 | |||
XM_011531214.2 | XP_011529516.1 | 2 | 806 | |||
XM_011531215.2 | XP_011529517.1 | 2 | 672 | |||
GenBank mRNA | AL549300.3 | - | 2 | 298 | ||
AY445048.1 | - | 1 | 25 | |||
BC001909.1 | - | 2 | 222 | 3537447 | ||
BC007232.1 | - | 2 | 222 | 3347302 | ||
BC032121.2 | - | 2 | 230 | 4575331 | ||
BC071851.1 | - | 2 | 83 | 5934127 | ||
BG766429.1 | - | 2 | 228 | |||
DQ323993.1 | - | 1 | 76 | |||
DT217835.1 | - | 2 | 238 |