Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 16 | 2022 | ||
XM_011545352.2 | XP_011543654.1 | 17 | 2191 | |||
XM_017018560.1 | XP_016874049.1 | 16 | 2055 | |||
GenBank mRNA | AF172822.1 | - | 3 | 408 | ||
AK290866.1 | - | 16 | 2018 | |||
AL834171.1 | - | 16 | 1965 | |||
BC006369.1 | - | 4 | 391 | |||
BC008013.1 | - | 12 | 1475 | |||
BC009859.2 | - | 6 | 681 | |||
BC015664.2 | - | 17 | 2332 | 4554197 |