Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTSS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138383.2 | NP_612392.1 | 11 | 1157 | ||
XM_011523450.2 | XP_011521752.1 | 12 | 1061 | |||
XM_011523453.2 | XP_011521755.2 | 11 | 1313 | |||
XM_011523454.2 | XP_011521756.2 | 10 | 1304 | |||
XM_011523458.2 | XP_011521760.1 | 5 | 538 | |||
XM_017023880.1 | XP_016879369.1 | 11 | 1313 | |||
XM_017023882.1 | XP_016879371.1 | 11 | 1313 | |||
XM_017023883.1 | XP_016879372.1 | 11 | 1313 | |||
XM_017023884.1 | XP_016879373.1 | 11 | 1443 | |||
GenBank mRNA | AB115770.1 | - | 11 | 897 |