Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319941.1 | NP_001306870.1 | 2 | 429 | ||
NM_203411.1 | NP_981956.1 | 2 | 755 | |||
XM_005256856.3 | XP_005256913.1 | 2 | 977 | |||
GenBank mRNA | AK000032.1 | - | 2 | 756 | ||
BC057812.1 | - | 2 | 755 | 30332216 |