Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138384.2 | NP_612393.2 | 3 | 306 | ||
GenBank mRNA | AK074976.1 | - | 3 | 227 | ||
AK125012.1 | - | 4 | 593 | |||
AK292515.1 | - | 3 | 148 | |||
AK293988.1 | - | 3 | 214 | |||
AK308997.1 | - | 3 | 255 | |||
BC000920.2 | - | 1 | 15 | |||
BC004409.1 | - | 3 | 265 | |||
BC026039.1 | - | 3 | 282 | 5444745 | ||
BC026725.1 | - | 3 | 185 | |||
BC035721.1 | - | 3 | 210 | 5588933 | ||
DB136242.1 | - | 3 | 306 |