Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033100.3 | NP_149091.1 | 17 | 2416 | ||
XM_011540337.1 | XP_011538639.1 | 16 | 2907 | |||
XM_011540339.1 | XP_011538641.1 | 17 | 2222 | |||
GenBank mRNA | AB053448.1 | - | 17 | 2289 | ||
AB593154.1 | - | 17 | 2416 | |||
AB593155.1 | - | 17 | 2402 | |||
AK124752.1 | - | 10 | 1505 | |||
AL080188.1 | - | 2 | 722 | |||
DA373888.1 | - | 1 | 265 |