Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171971.2 | NP_001165442.1 | 16 | 2142 | ||
NM_033100.3 | NP_149091.1 | 16 | 2142 | |||
XM_011540337.1 | XP_011538639.1 | 15 | 2633 | |||
XM_011540338.1 | XP_011538640.1 | 15 | 2633 | |||
XM_011540339.1 | XP_011538641.1 | 16 | 1948 | |||
GenBank mRNA | AB053448.1 | - | 16 | 2015 | ||
AB593154.1 | - | 16 | 2142 | |||
AB593155.1 | - | 16 | 2128 | |||
AK124752.1 | - | 9 | 1231 | |||
AL080188.1 | - | 1 | 448 | |||
BC038799.1 | - | 16 | 2132 | 5492127 |