Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322923.1 | NP_001309852.1 | 5 | 731 | ||
NM_001322924.1 | NP_001309853.1 | 4 | 659 | |||
NM_052862.3 | NP_443094.3 | 6 | 777 | |||
GenBank mRNA | AF545852.1 | - | 6 | 921 | ||
AK025425.1 | - | 1 | 116 | |||
AL137762.1 | - | 6 | 698 | |||
AL833211.1 | - | 4 | 467 | |||
BC072399.1 | - | 1 | 69 | |||
BC101536.1 | - | 6 | 614 | 8069042 | ||
BC101562.1 | - | 6 | 614 | 8069068 | ||
BC143428.1 | - | 5 | 524 | 9051936 |