Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144575.2 | NP_653176.2 | 21 | 2144 | ||
XM_011533159.2 | XP_011531461.1 | 21 | 2141 | |||
XM_011533160.2 | XP_011531462.1 | 22 | 2332 | |||
XM_011533161.2 | XP_011531463.1 | 21 | 2141 | |||
XM_017005264.1 | XP_016860753.1 | 22 | 2332 | |||
XM_017005265.1 | XP_016860754.1 | 21 | 2141 | |||
XM_017005266.1 | XP_016860755.1 | 20 | 2129 | |||
GenBank mRNA | AK026692.1 | - | 5 | 1159 | ||
AK027176.1 | - | 9 | 1793 | |||
AK290298.1 | - | 21 | 2145 | |||
BC117345.1 | - | 21 | 2105 | 40125896 | ||
BC117347.1 | - | 21 | 2105 | 40125898 | ||
BX640793.1 | - | 21 | 2144 | |||
BX647678.1 | - | 14 | 2370 |