Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 6 | 749 | ||
NM_001025433.1 | NP_001020604.1 | 5 | 647 | |||
NM_001025434.1 | NP_001020605.1 | 5 | 635 | |||
NM_001286137.1 | NP_001273066.1 | 4 | 571 | |||
GenBank mRNA | AK223050.1 | - | 6 | 661 | ||
AK297125.1 | - | 4 | 463 | |||
AK298896.1 | - | 6 | 497 | |||
AK307533.1 | - | 6 | 611 | |||
AK312368.1 | - | 6 | 675 | |||
AK316246.1 | - | 5 | 470 | |||
BC007659.2 | - | 6 | 675 | 3349257 | ||
BC107739.1 | - | 5 | 569 | |||
BE903613.1 | - | 6 | 697 | |||
CD014005.1 | - | 5 | 569 | |||
J03934.1 | - | 6 | 608 |