Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003786.2 | NP_001003786.1 | 7, 8 | 924 | ||
NM_001003787.2 | NP_001003787.1 | 9, 10 | 1035 | |||
NM_001003788.2 | NP_001003788.1 | 8, 9 | 1006 | |||
NM_001165969.1 | NP_001159441.1 | 7, 8 | 948 | |||
NM_001165970.1 | NP_001159442.1 | 7, 8 | 903 | |||
NM_153335.5 | NP_699166.2 | 7, 8 | 924 | |||
XM_005257797.2 | XP_005257854.1 | 9, 10 | 886 | |||
XM_005257798.2 | XP_005257855.1 | 7, 8 | 848 | |||
XM_005257799.2 | XP_005257856.1 | 8, 9 | 857 | |||
XM_005257800.2 | XP_005257857.1 | 9, 10 | 910 | |||
XM_005257801.4 | XP_005257858.1 | 9, 10 | 910 | |||
XM_005257803.4 | XP_005257860.1 | 8, 9 | 738 | |||
XM_011525466.2 | XP_011523768.1 | 9, 10 | 910 | |||
XM_011525467.2 | XP_011523769.1 | 9, 10 | 937 | |||
XM_017025312.1 | XP_016880801.1 | 7, 8 | 799 | |||
XM_017025313.1 | XP_016880802.1 | 8, 9 | 881 | |||
XM_017025314.1 | XP_016880803.1 | 7, 8 | 848 | |||
GenBank mRNA | AF308302.1 | - | 9, 10 | 874 | ||
AK074771.1 | - | 7, 8 | 790 | |||
AK075005.1 | - | 7, 8 | 753 | |||
AK293160.1 | - | 7, 8 | 811 | |||
AK301331.1 | - | 7, 8 | 775 | |||
AL832407.1 | - | 8, 9 | 819 | |||
AY290821.2 | - | 7, 8 | 924 |