Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146039.1 | NP_001139511.1 | 2 | 455 | ||
NM_001320252.1 | NP_001307181.1 | 4 | 755 | |||
NM_152281.2 | NP_689494.2 | 2 | 455 | |||
NR_027397.1 | 2 | 455 | ||||
XM_006711628.3 | XP_006711691.1 | 4 | 726 | |||
XM_011510149.2 | XP_011508451.1 | 2 | 392 | |||
XM_011510150.2 | XP_011508452.1 | 3 | 629 | |||
XM_011510151.1 | XP_011508453.1 | 2 | 378 | |||
XM_017002807.1 | XP_016858296.1 | 4 | 692 | |||
GenBank mRNA | AK021814.1 | - | 2 | 448 | ||
AK308339.1 | - | 2 | 455 | |||
BC047476.1 | - | 4 | 726 | 5296051 | ||
BC064945.1 | - | 2 | 446 | 6019822 |