Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173512.1 | NP_001166983.1 | 4 | 696 | ||
NM_138777.3 | NP_620132.1 | 5 | 852 | |||
NM_199177.2 | NP_954646.1 | 4 | 516 | |||
XM_005252302.3 | XP_005252359.1 | 6 | 920 | |||
XM_011519183.2 | XP_011517485.1 | 6 | 1175 | |||
XM_011519184.2 | XP_011517486.1 | 6 | 685 | |||
XM_011519185.2 | XP_011517487.1 | 6 | 920 | |||
XM_011519186.1 | XP_011517488.1 | 5 | 600 | |||
XM_011519187.2 | XP_011517489.1 | 4 | 640 | |||
GenBank mRNA | AK291603.1 | - | 5 | 542 | ||
AK291809.1 | - | 7 | 876 | |||
AK292790.1 | - | 7 | 876 | |||
AK298046.1 | - | 6 | 1358 | |||
AK300718.1 | - | 4 | 399 | |||
BC013049.1 | - | 4 | 513 | 3882448 | ||
BC053604.1 | - | 4 | 514 | 6008684 | ||
BF982227.1 | - | 587 |