Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173512.1 | NP_001166983.1 | 2 | 408 | ||
NM_138777.3 | NP_620132.1 | 2 | 408 | |||
NM_199177.2 | NP_954646.1 | 1 | 72 | |||
XM_005252302.3 | XP_005252359.1 | 3 | 476 | |||
XM_011519183.2 | XP_011517485.1 | 3 | 731 | |||
XM_011519184.2 | XP_011517486.1 | 3 | 241 | |||
XM_011519185.2 | XP_011517487.1 | 3 | 476 | |||
XM_011519186.1 | XP_011517488.1 | 2 | 156 | |||
XM_017015286.1 | XP_016870775.1 | 3 | 476 | |||
XM_017015287.1 | XP_016870776.1 | 2 | 156 | |||
GenBank mRNA | AK291603.1 | - | 2 | 98 | ||
AK291809.1 | - | 2 | 137 | |||
AK292790.1 | - | 2 | 137 | |||
AK298046.1 | - | 2 | 127 | |||
AK300718.1 | - | 2 | 111 | |||
AK316266.1 | - | 3 | 425 | |||
BC013049.1 | - | 1 | 69 | 3882448 | ||
BC053604.1 | - | 1 | 70 | 6008684 | ||
BF982227.1 | - | 143 |