Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHAL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033195.2 | NP_149972.1 | 1 | 792 | ||
GenBank mRNA | AK058192.1 | - | 1 | 722 | ||
AK312777.1 | - | 1 | 742 | |||
AY009108.1 | - | 1 | 765 | |||
AY642121.1 | - | 1 | 785 | |||
BC022034.1 | - | 1 | 778 | 4825988 |