Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPECC1 SIRNA Assays ›
Gene Symbol:
SPECC1
Gene Name:
sperm antigen with calponin homology and coiled-coil domains 1
Gene Aliases:
CYTSB, HCMOGT-1, HCMOGT1, NSP
Chromosome Location:
Chr.17: 20009301 - 20319026 on Build GRCh38
UniGene ID:
Hs.431045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40961
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPECC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033553.2
NP_001028725.1
5
1949
NM_001033554.2
NP_001028726.1
3
1858
NM_001033555.2
NP_001028727.1
3
1858
NM_001243438.1
NP_001230367.1
3
1858
NM_001243439.1
NP_001230368.1
5
2008
NM_152904.4
NP_690868.3
5
1949
XM_005256860.2
XP_005256917.1
2
278
XM_011524075.1
XP_011522377.1
4
1901
XM_017025317.1
XP_016880806.1
4
1925
XM_017025318.1
XP_016880807.1
4
1925
XM_017025319.1
XP_016880808.1
5
2067
XM_017025320.1
XP_016880809.1
5
2043
XM_017025321.1
XP_016880810.1
3
1882
XM_017025322.1
XP_016880811.1
4
1925
XM_017025323.1
XP_016880812.1
4
1925
XM_017025324.1
XP_016880813.1
4
1925
XM_017025325.1
XP_016880814.1
5
2043
GenBank mRNA
AB041533.1
-
5
1949
AK295093.1
-
5
1981
AK295811.1
-
2
1485
AK298601.1
-
2
690
AK301318.1
-
2
200
AK304186.1
-
3
1780
AK314341.1
-
5
2024
AY816326.1
-
3
1763
AY816327.1
-
3
1763
AY816328.1
-
5
1949
AY816329.1
-
5
1949
BC021123.1
-
3
1756
3614358
BC033618.1
-
2
973
BC050058.1
-
3
1693
5277450
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-fr72p:80/10.239.161.22:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0