Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052838.4 | NP_443070.5 | 2, 3 | 188 | ||
XM_017022991.1 | XP_016878480.1 | 2, 3 | 168 | |||
GenBank mRNA | AF085235.1 | - | 1, 2 | 22 | ||
AK301045.1 | - | 1, 2 | 20 | |||
AK303390.1 | - | 2, 3 | 151 | |||
AY034176.1 | - | 1, 2 | 22 | |||
BQ072902.1 | - | 2, 3 | 201 |