Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138389.2 | NP_612398.2 | 15 | 1863 | ||
NR_033290.1 | 13 | 1358 | ||||
XM_005262672.2 | XP_005262729.1 | 14 | 1669 | |||
XM_006714033.2 | XP_006714096.1 | 14 | 1434 | |||
GenBank mRNA | AK094179.1 | - | 7 | 613 | ||
AK292831.1 | - | 15 | 1790 | |||
AK296859.1 | - | 11 | 1045 | |||
AK304050.1 | - | 13 | 1340 | |||
AK308959.1 | - | 6 | 678 | |||
BC001096.2 | - | 13 | 1614 | 3507281 | ||
BC040452.1 | - | 15 | 1780 | 3846822 | ||
BX640862.1 | - | 12 | 1099 |