Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138389.2 | NP_612398.2 | 7 | 1009 | ||
NR_033290.1 | 5 | 504 | ||||
XM_005262672.2 | XP_005262729.1 | 6 | 815 | |||
XM_006714033.2 | XP_006714096.1 | 6 | 580 | |||
XM_017008838.1 | XP_016864327.1 | 6 | 815 | |||
GenBank mRNA | AK292831.1 | - | 7 | 936 | ||
AK296859.1 | - | 3 | 191 | |||
AK304050.1 | - | 5 | 486 | |||
BC001096.2 | - | 5 | 760 | 3507281 | ||
BC040452.1 | - | 7 | 926 | 3846822 | ||
BX640862.1 | - | 4 | 245 |