Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPH7 SIRNA Assays ›
Gene Symbol:
DPH7
Gene Name:
diphthamide biosynthesis 7
Gene Aliases:
C9orf112, RRT2, WDR85
Chromosome Location:
Chr.9: 137554899 - 137578935 on Build GRCh38
UniGene ID:
Hs.292570
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41006
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138778.3
NP_620133.1
5
658
XM_005266123.2
XP_005266180.1
6
1116
XM_005266126.3
XP_005266183.1
4
889
XM_005266128.2
XP_005266185.1
6
742
XM_005266129.2
XP_005266186.1
6
745
XM_005266130.2
XP_005266187.1
4
513
XM_011519188.1
XP_011517490.1
5
647
XM_011519189.1
XP_011517491.1
5
759
XM_011519190.2
XP_011517492.1
5
647
XM_011519191.2
XP_011517493.1
5
647
XM_011519192.2
XP_011517494.1
5
647
XM_011519193.2
XP_011517495.1
5
647
XM_011519194.1
XP_011517496.1
5
647
XM_011519199.2
XP_011517501.1
5
604
XM_011519200.1
XP_011517502.1
5
607
XM_017015289.1
XP_016870778.1
5
647
XM_017015290.1
XP_016870779.1
5
647
XM_017015291.1
XP_016870780.1
6
951
XM_017015292.1
XP_016870781.1
5
1021
XM_017015293.1
XP_016870782.1
5
978
XM_017015294.1
XP_016870783.1
7
931
XM_017015295.1
XP_016870784.1
5
1018
XM_017015296.1
XP_016870785.1
5
857
XM_017015297.1
XP_016870786.1
7
928
XM_017015298.1
XP_016870787.1
6
1211
XM_017015299.1
XP_016870788.1
6
468
XM_017015300.1
XP_016870789.1
6
1208
XM_017015301.1
XP_016870790.1
4
881
XM_017015302.1
XP_016870791.1
4
1935
XM_017015303.1
XP_016870792.1
5
1116
XM_017015304.1
XP_016870793.1
4
581
XM_017015305.1
XP_016870794.1
5
1978
XM_017015306.1
XP_016870795.1
6
1116
XM_017015307.1
XP_016870796.1
6
1113
XM_017015308.1
XP_016870797.1
4
1840
XM_017015309.1
XP_016870798.1
6
840
GenBank mRNA
AK075115.1
-
5
659
AK307180.1
-
4
837
AK308998.1
-
4
513
BC003123.1
-
5
597
3502107
BC017335.2
-
5
617
4642600
Back To Top
More Information
Panther Classification:
Molecular Function -
methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
diphthine methylesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0