Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033197.2 | NP_149974.2 | 5 | 666 | ||
GenBank mRNA | AF364078.1 | - | 5 | 591 | ||
AK290243.1 | - | 5 | 648 | |||
AK310964.1 | - | 5 | 590 | |||
AK313176.1 | - | 5 | 584 | |||
BC008429.1 | - | 5 | 581 | 4291561 |