Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033197.2 | NP_149974.2 | 12 | 1349 | ||
GenBank mRNA | AF364078.1 | - | 12 | 1274 | ||
AK127971.1 | - | 9 | 848 | |||
AK290243.1 | - | 12 | 1331 | |||
AK313176.1 | - | 12 | 1267 | |||
AY358595.1 | - | 12 | 1261 | |||
BC008429.1 | - | 12 | 1264 | 4291561 |