Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033647.4 | NP_387467.2 | 2 | 507 | ||
NR_135080.1 | 2 | 507 | ||||
NR_135081.1 | 2 | 507 | ||||
XM_005269234.2 | XP_005269291.1 | 2 | 485 | |||
XM_017020230.1 | XP_016875719.1 | 2 | 485 | |||
GenBank mRNA | AF319995.1 | - | 2 | 448 | ||
AK290894.1 | - | 2 | 472 | |||
BC143409.1 | - | 2 | 473 | |||
BC143411.1 | - | 2 | 473 | |||
DN997712.1 | - | 2 | 516 |