Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138392.3 | NP_612401.2 | 6 | 405 | ||
XM_006723474.1 | XP_006723537.1 | 6 | 405 | |||
XM_006723475.1 | XP_006723538.1 | 6 | 405 | |||
XM_011527492.1 | XP_011525794.1 | 6 | 405 | |||
XM_011527493.1 | XP_011525795.1 | 5 | 301 | |||
XM_017027473.1 | XP_016882962.1 | 6 | 405 | |||
XM_017027474.1 | XP_016882963.1 | 5 | 301 | |||
GenBank mRNA | AF258553.1 | - | 6 | 404 | ||
AK075057.1 | - | 6 | 401 | |||
AK123695.1 | - | 4 | 383 | |||
AK300808.1 | - | 4 | 284 | |||
AK312748.1 | - | 6 | 369 | |||
BC022855.1 | - | 6 | 376 | 4102901 | ||
DB038927.1 | - | 6 | 405 |