Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138396.5 | NP_612405.2 | 3 | 966 | ||
GenBank mRNA | AK313925.1 | - | 1 | 643 | ||
BC009489.2 | - | 1 | 446 | 3936171 | ||
BC036455.2 | - | 3 | 947 | 4811410 | ||
BC050397.1 | - | 3 | 966 |