Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286775.1 | NP_001273704.1 | 3 | 390 | ||
NM_001286776.1 | NP_001273705.1 | 2 | 315 | |||
NM_138779.4 | NP_620134.3 | 3 | 390 | |||
NR_104590.1 | 2 | 315 | ||||
XM_005254097.2 | XP_005254154.1 | 3 | 444 | |||
XM_011521135.2 | XP_011519437.1 | 3 | 1031 | |||
XM_017020852.1 | XP_016876341.1 | 3 | 1375 | |||
GenBank mRNA | AF070559.1 | - | 1 | 904 | ||
AK314995.1 | - | 3 | 319 | |||
BC015148.2 | - | 3 | 361 | 4825712 | ||
BC048011.1 | - | 1 | 4423 | |||
BG527702.1 | - | 3 | 339 | |||
BI559423.1 | - | 2 | 288 | |||
BI861260.1 | - | 2 | 249 | |||
BP363423.1 | - | 2 | 293 | |||
HY045638.1 | - | 2 | 315 |