Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035516.3 | NP_001030593.1 | 2, 3 | 140 | ||
NM_001126056.2 | NP_001119528.2 | 14, 15 | 1440 | |||
NM_001126059.2 | NP_001119531.1 | 8, 9 | 491 | |||
NM_001308383.1 | NP_001295312.1 | 7, 8 | 461 | |||
NM_033317.4 | NP_201574.3 | 12, 13 | 1473 | |||
GenBank mRNA | AF086315.1 | - | 2, 3 | 131 | ||
AL832080.1 | - | 2, 3 | 141 | |||
AY622964.1 | - | 2, 3 | 126 | |||
AY622965.1 | - | 12, 13 | 1290 | |||
AY789695.1 | - | 12, 13 | 1296 | |||
AY789698.1 | - | 10, 11 | 525 | |||
AY789700.1 | - | 9, 10 | 465 | |||
AY789701.1 | - | 11, 12 | 681 | |||
AY789704.1 | - | 9, 10 | 489 | |||
AY789705.1 | - | 8, 9 | 429 | |||
AY789706.1 | - | 7, 8 | 378 | |||
AY789707.1 | - | 2, 3 | 129 | |||
BC035311.1 | - | 12, 13 | 1467 | 4752921 | ||
BP326188.1 | - | 8, 9 | 490 | |||
CN369399.1 | - | 7, 8 | 400 | |||
EF158320.1 | - | 14, 15 | 1267 |