Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136239.1 | NP_001129711.1 | 4, 5 | 1415 | ||
XM_011543726.2 | XP_011542028.1 | 3, 4 | 1478 | |||
GenBank mRNA | AF272898.1 | - | 4, 5 | 1065 | ||
AK297992.1 | - | 3, 4 | 1031 |