Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152290.3 | NP_689503.3 | 3 | 540 | ||
XM_011542434.1 | XP_011540736.1 | 2 | 411 | |||
GenBank mRNA | AK026705.1 | - | 3 | 508 | ||
AK298760.1 | - | 3 | 502 | |||
BC029894.1 | - | 3 | 511 | 5171302 |