Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144577.3 | NP_653178.3 | 14 | 2963 | ||
XM_005259413.3 | XP_005259470.1 | 16 | 2671 | |||
XM_005259414.3 | XP_005259471.1 | 16 | 2667 | |||
XM_005259415.3 | XP_005259472.1 | 16 | 2685 | |||
XM_005259416.4 | XP_005259473.1 | 9 | 1818 | |||
XM_011527515.2 | XP_011525817.1 | 17 | 3614 | |||
XM_011527516.2 | XP_011525818.1 | 15 | 2404 | |||
XM_017027483.1 | XP_016882972.1 | 15 | 3444 | |||
GenBank mRNA | AK057357.1 | - | 12 | 1671 | ||
AK057488.1 | - | 8 | 1661 | |||
AK128144.1 | - | 15 | 3027 | |||
AL122083.1 | - | 7 | 1653 | |||
BC007730.2 | - | 14 | 2963 | |||
BC025752.1 | - | 13 | 2480 |