Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143763.1 | NP_001137235.1 | 4, 5 | 383 | ||
NM_001143764.1 | NP_001137236.1 | 4, 5 | 383 | |||
NM_130784.2 | NP_570140.1 | 4, 5 | 306 | |||
GenBank mRNA | AK314978.1 | - | 4, 5 | 333 | ||
AY027807.1 | - | 4, 5 | 306 | |||
AY027808.1 | - | 1, 2 | 1242 | |||
BC034821.1 | - | 4, 5 | 333 | 5164386 | ||
DB053356.1 | - | 4, 5 | 383 |